Assay Design



Gene Transcripts

Gene Symbol
KRTAP3-3
Entrez Gene ID
85293
Gene Name
keratin associated protein 3-3
Gene Aliases
KAP3.3, KRTAP3.3
Chromosome Location
Chr.17: 40993430 - 40994133 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033185.2 NP_149441.1 1 - 1 527 104
GenBank mRNA AJ406933.1 - 1 - 1 527 104
BC069099.1 - 1 - 1 527 104

View Full Product Details