Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172357.1 | NP_001165828.1 | 4 - 5 | 595 | 87 |
NM_001317778.1 | NP_001304707.1 | 4 - 5 | 595 | 87 | |
NM_001317780.1 | NP_001304709.1 | 4 - 5 | 595 | 87 | |
GenBank mRNA | BF433929.1 | - | 2 - 3 | 87 | |
BG535994.1 | - | 4 - 5 | 472 | 87 | |
BG569816.1 | - | 4 - 5 | 472 | 87 | |
BI524154.1 | - | 4 - 5 | 451 | 87 | |
DQ884411.1 | - | 4 - 5 | 437 | 87 | |
KU178330.1 | - | 4 - 5 | 437 | 87 |