Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282710.1 | NP_001269639.1 | 1 - 2 | 316 | 62 |
NM_001282711.1 | NP_001269640.1 | 1 - 2 | 316 | 62 | |
NM_001282712.1 | NP_001269641.1 | 1 - 2 | 316 | 62 | |
NM_152391.4 | NP_689604.1 | 1 - 2 | 316 | 62 | |
NR_104231.1 | 1 - 2 | 316 | 62 | ||
XM_011510315.1 | XP_011508617.1 | 1 - 2 | 316 | 62 | |
GenBank mRNA | AK001091.2 | - | 1 - 2 | 232 | 62 |
AK301439.1 | - | 1 - 2 | 211 | 62 | |
AK313402.1 | - | 1 - 2 | 211 | 62 | |
AL548337.3 | - | 1 - 2 | 87 | 62 | |
BC027625.1 | - | 1 - 2 | 280 | 62 | |
BI601351.1 | - | 1 - 2 | 262 | 62 | |
BQ277359.1 | - | 1 - 2 | 80 | 62 | |
CX751769.1 | - | 1 - 2 | 153 | 62 | |
DB460796.1 | - | 1 - 2 | 316 | 62 | |
DC392668.1 | - | 1 - 2 | 300 | 62 |