Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128301.2 | NP_001121773.1 | 4 - 5 | 705 | 133 |
NM_001128302.2 | NP_001121774.1 | 3 - 4 | 473 | 133 | |
NM_001302835.1 | NP_001289764.1 | 5 - 6 | 649 | 133 | |
NM_001302836.1 | NP_001289765.1 | 5 - 6 | 801 | 133 | |
NM_020424.4 | NP_065157.1 | 4 - 5 | 624 | 133 | |
NR_126516.1 | 3 - 4 | 546 | 133 | ||
NR_126520.1 | 2 - 3 | 328 | 133 | ||
NR_126521.1 | 2 - 3 | 314 | 133 | ||
NR_126527.1 | 2 - 3 | 142 | 133 | ||
NR_126528.1 | 3 - 4 | 560 | 133 | ||
XM_005255444.1 | XP_005255501.1 | 4 - 5 | 526 | 133 | |
XM_011545900.2 | XP_011544202.1 | 4 - 5 | 288 | 133 | |
XM_011545901.2 | XP_011544203.1 | 4 - 5 | 354 | 133 | |
XM_011545902.1 | XP_011544204.1 | 5 - 6 | 625 | 133 | |
XM_011545908.2 | XP_011544210.1 | 4 - 5 | 356 | 133 | |
XM_017023475.1 | XP_016878964.1 | 5 - 6 | 1041 | 133 | |
XM_017023477.1 | XP_016878966.1 | 3 - 4 | 876 | 133 | |
XM_017023478.1 | XP_016878967.1 | 4 - 5 | 346 | 133 | |
XM_017023479.1 | XP_016878968.1 | 4 - 5 | 783 | 133 | |
XM_017023480.1 | XP_016878969.1 | 6 - 7 | 869 | 133 | |
XM_017023481.1 | XP_016878970.1 | 4 - 5 | 498 | 133 | |
XM_017023482.1 | XP_016878971.1 | 3 - 4 | 552 | 133 | |
GenBank mRNA | AK308690.1 | - | 2 - 3 | 393 | 133 |
AK311415.1 | - | 3 - 4 | 548 | 133 | |
AK311881.1 | - | 3 - 4 | 340 | 133 | |
AV714321.1 | - | 3 - 4 | 319 | 133 | |
BC017039.1 | - | 5 - 6 | 649 | 133 | |
BG541469.1 | - | 2 - 3 | 177 | 133 | |
BI825882.1 | - | 5 - 6 | 648 | 133 | |
BM546065.1 | - | 2 - 3 | 278 | 133 | |
BX096767.1 | - | 2 - 3 | 142 | 133 | |
CD237780.1 | - | 3 - 4 | 343 | 133 | |
CX163936.1 | - | 3 - 4 | 350 | 133 | |
DA572182.1 | - | 4 - 5 | 590 | 133 |