Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018240.6 | NP_060710.3 | 2 - 3 | 606 | 123 |
XM_005245305.4 | XP_005245362.1 | 2 - 3 | 263 | 123 | |
GenBank mRNA | AY017369.1 | - | 2 - 3 | 202 | 123 |
AY302131.1 | - | 2 - 3 | 610 | 123 | |
BC109192.1 | - | 2 - 3 | 269 | 123 | |
BC109193.1 | - | 2 - 3 | 269 | 123 |