Assay Design
Gene Transcripts
- Gene Symbol
- NFYA
- Entrez Gene ID
-
4800
- Gene Name
- nuclear transcription factor Y subunit alpha
- Gene Aliases
-
CBF-A, CBF-B, HAP2, NF-YA
- Chromosome Location
-
Chr.6: 41072968 - 41102407 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_002505.4 |
NP_002496.1 |
2 - 3 |
258 |
104 |
GenBank mRNA |
X59711.1 |
- |
2 - 3 |
254 |
104 |
View Full Product Details