Assay Design



Gene Transcripts

Gene Symbol
CENPQ
Entrez Gene ID
55166
Gene Name
centromere protein Q
Gene Aliases
C6orf139, CENP-Q
Chromosome Location
Chr.6: 49463327 - 49493107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005249205.1 XP_005249262.1 1 - 2 110 93

View Full Product Details