Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019105.6 | NP_061978.6 | 31 - 32 | 10805 | 150 |
GenBank mRNA | AB209012.1 | - | 2 - 3 | 531 | 150 |
BC033740.1 | - | 2 - 3 | 382 | 150 | |
BC071883.1 | - | 1 - 2 | 436 | 150 | |
M25813.1 | - | 2 - 3 | 337 | 150 |