Assay Design



Gene Transcripts

Gene Symbol
TNXB
Entrez Gene ID
7148
Gene Name
tenascin XB
Gene Aliases
EDS3, HXBL, TENX, TN-X, TNX, TNXB1, TNXB2, TNXBS, VUR8, XB, XBS
Chromosome Location
Chr.6: 32041155 - 32109374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019105.6 NP_061978.6 9 - 10 3937 95

View Full Product Details