Assay Design
Gene Transcripts
- Gene Symbol
- TNXB
- Entrez Gene ID
-
7148
- Gene Name
- tenascin XB
- Gene Aliases
-
EDS3, HXBL, TENX, TN-X, TNX, TNXB1, TNXB2, TNXBS, VUR8, XB, XBS
- Chromosome Location
-
Chr.6: 32041155 - 32109374 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_019105.6 |
NP_061978.6 |
14 - 15 |
5480 |
71 |
GenBank mRNA |
Y13782.1 |
- |
2 - 3 |
466 |
71 |
Y17866.1 |
- |
5 - 6 |
1294 |
71 |
View Full Product Details