Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005557.3 | NP_005548.2 | 2 - 3 | 750 | 100 |
GenBank mRNA | AF061812.1 | - | 2 - 3 | 625 | 100 |
AK290853.1 | - | 2 - 3 | 688 | 100 | |
BC039169.1 | - | 2 - 3 | 661 | 100 | |
S79867.1 | - | 2 - 3 | 611 | 100 |