Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007111.4 | NP_009042.1 | 2 - 3 | 226 | 64 |
NR_026580.1 | 2 - 3 | 121 | 64 | ||
XM_005268327.1 | XP_005268384.1 | 2 - 3 | 111 | 64 | |
XM_005268328.2 | XP_005268385.1 | 2 - 3 | 164 | 64 | |
XM_005268331.1 | XP_005268388.1 | 2 - 3 | 228 | 64 | |
XM_011537518.2 | XP_011535820.1 | 1 - 2 | 78 | 64 | |
XM_011537519.1 | XP_011535821.1 | 2 - 3 | 228 | 64 | |
XM_011537520.2 | XP_011535822.1 | 2 - 3 | 111 | 64 | |
XM_017020717.1 | XP_016876206.1 | 2 - 3 | 111 | 64 | |
XM_017020718.1 | XP_016876207.1 | 2 - 3 | 111 | 64 | |
XM_017020719.1 | XP_016876208.1 | 2 - 3 | 111 | 64 | |
XM_017020720.1 | XP_016876209.1 | 2 - 3 | 194 | 64 | |
XM_017020721.1 | XP_016876210.1 | 2 - 3 | 116 | 64 | |
XM_017020722.1 | XP_016876211.1 | 2 - 3 | 111 | 64 | |
GenBank mRNA | AK297114.1 | - | 2 - 3 | 111 | 64 |
AK298521.1 | - | 2 - 3 | 233 | 64 | |
AK310401.1 | - | 2 - 3 | 108 | 64 | |
AK313742.1 | - | 1 - 2 | 72 | 64 | |
BC011685.2 | - | 2 - 3 | 226 | 64 | |
CD623680.1 | - | 2 - 3 | 121 | 64 | |
L23959.1 | - | 1 - 2 | 51 | 64 |