Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015205.2 | NP_056020.2 | 25 - 26 | 3083 | 103 |
NM_032189.3 | NP_115565.3 | 25 - 26 | 3083 | 103 | |
XM_005268299.4 | XP_005268356.1 | 25 - 26 | 3378 | 103 | |
XM_005268300.4 | XP_005268357.1 | 25 - 26 | 3377 | 103 | |
XM_005268301.4 | XP_005268358.1 | 25 - 26 | 3378 | 103 | |
XM_005268303.4 | XP_005268360.1 | 25 - 26 | 3378 | 103 | |
XM_005268305.4 | XP_005268362.1 | 25 - 26 | 3377 | 103 | |
XM_005268306.4 | XP_005268363.1 | 25 - 26 | 3377 | 103 | |
XM_011537480.2 | XP_011535782.1 | 25 - 26 | 3378 | 103 | |
XM_017020490.1 | XP_016875979.1 | 25 - 26 | 3378 | 103 | |
XM_017020491.1 | XP_016875980.1 | 25 - 26 | 3378 | 103 | |
XM_017020492.1 | XP_016875981.1 | 25 - 26 | 3377 | 103 | |
GenBank mRNA | AB028944.2 | - | 24 - 25 | 2900 | 103 |
AL161996.1 | - | 8 - 9 | 1129 | 103 |