Assay Design



Gene Transcripts

Gene Symbol
PLXNC1
Entrez Gene ID
10154
Gene Name
plexin C1
Gene Aliases
CD232, PLXN-C1, VESPR
Chromosome Location
Chr.12: 94148696 - 94307675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005761.2 NP_005752.1 18 - 19 3369 141
XM_011537730.2 XP_011536032.1 18 - 19 3396 141
XM_017018671.1 XP_016874160.1 18 - 19 3396 141
XM_017018672.1 XP_016874161.1 18 - 19 3396 141
GenBank mRNA AB208934.1 - 17 - 18 2050 141
AF030339.1 - 18 - 19 3369 141
AK295226.1 - 3 - 4 498 141
AK308985.1 - 4 - 5 302 141

View Full Product Details