Assay Design



Gene Transcripts

Gene Symbol
PLXNC1
Entrez Gene ID
10154
Gene Name
plexin C1
Gene Aliases
CD232, PLXN-C1, VESPR
Chromosome Location
Chr.12: 94148696 - 94307675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005761.2 NP_005752.1 25 - 26 4214 74
NR_037687.1 6 - 7 758 74
XM_006719186.3 XP_006719249.1 24 - 25 4202 74
GenBank mRNA AB208934.1 - 24 - 25 2895 74
AF030339.1 - 25 - 26 4214 74
AK295226.1 - 10 - 11 1343 74
AK304327.1 - 6 - 7 758 74
AK307920.1 - 6 - 7 661 74
AK308985.1 - 11 - 12 1147 74

View Full Product Details