Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005761.2 | NP_005752.1 | 2 - 3 | 1450 | 156 |
XM_006719186.3 | XP_006719249.1 | 2 - 3 | 1477 | 156 | |
XM_011537730.2 | XP_011536032.1 | 2 - 3 | 1477 | 156 | |
XM_011537731.2 | XP_011536033.1 | 2 - 3 | 1477 | 156 | |
XM_017018671.1 | XP_016874160.1 | 2 - 3 | 1477 | 156 | |
XM_017018672.1 | XP_016874161.1 | 2 - 3 | 1477 | 156 | |
GenBank mRNA | AB208934.1 | - | 1 - 2 | 131 | 156 |
AF030339.1 | - | 2 - 3 | 1450 | 156 |