Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005761.2 | NP_005752.1 | 9 - 10 | 2224 | 82 |
XM_006719186.3 | XP_006719249.1 | 9 - 10 | 2251 | 82 | |
XM_011537730.2 | XP_011536032.1 | 9 - 10 | 2251 | 82 | |
XM_011537731.2 | XP_011536033.1 | 9 - 10 | 2251 | 82 | |
XM_017018671.1 | XP_016874160.1 | 9 - 10 | 2251 | 82 | |
XM_017018672.1 | XP_016874161.1 | 9 - 10 | 2251 | 82 | |
GenBank mRNA | AF030339.1 | - | 9 - 10 | 2224 | 82 |