Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015267.3 | NP_056082.2 | 11 - 12 | 1203 | 88 |
XM_011538061.2 | XP_011536363.1 | 11 - 12 | 2713 | 88 | |
XM_011538063.2 | XP_011536365.1 | 10 - 11 | 2578 | 88 | |
XM_011538069.2 | XP_011536371.1 | 11 - 12 | 1052 | 88 | |
XM_011538070.2 | XP_011536372.1 | 11 - 12 | 1962 | 88 | |
XM_017019080.1 | XP_016874569.1 | 12 - 13 | 2045 | 88 | |
XM_017019081.1 | XP_016874570.1 | 12 - 13 | 1108 | 88 | |
GenBank mRNA | AB006631.2 | - | 11 - 12 | 1202 | 88 |
BC151245.1 | - | 11 - 12 | 1213 | 88 |