Assay Design



Gene Transcripts

Gene Symbol
CUX2
Entrez Gene ID
23316
Gene Name
cut like homeobox 2
Gene Aliases
CDP2, CUTL2
Chromosome Location
Chr.12: 111034024 - 111350556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015267.3 NP_056082.2 12 - 13 1263 126
XM_011538061.2 XP_011536363.1 12 - 13 2773 126
XM_011538063.2 XP_011536365.1 11 - 12 2638 126
XM_011538069.2 XP_011536371.1 12 - 13 1112 126
XM_011538070.2 XP_011536372.1 12 - 13 2022 126
XM_017019080.1 XP_016874569.1 13 - 14 2105 126
XM_017019081.1 XP_016874570.1 13 - 14 1168 126
GenBank mRNA AB006631.2 - 12 - 13 1262 126
BC151245.1 - 12 - 13 1273 126

View Full Product Details