Assay Design



Gene Transcripts

Gene Symbol
RBM38
Entrez Gene ID
55544
Gene Name
RNA binding motif protein 38
Gene Aliases
HSRNASEB, RNPC1, SEB4B, SEB4D, dJ800J21.2
Chromosome Location
Chr.20: 57391398 - 57409333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017495.5 NP_059965.2 1 - 2 422 79
NM_183425.2 NP_906270.1 1 - 2 422 79
GenBank mRNA AF432218.1 - 1 - 2 418 79
BC018711.1 - 1 - 2 174 79
BM549757.1 - - 417 79
DB504051.1 - 1 - 2 423 79
DN998561.1 - 1 - 2 407 79
X75314.1 - 1 - 2 209 79

View Full Product Details