Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004004.5 | NP_003995.2 | 1 - 2 | 194 | 104 |
GenBank mRNA | AK225966.1 | - | 1 - 2 | 157 | 104 |
AK313595.1 | - | 1 - 2 | 186 | 104 | |
BC017048.1 | - | 1 - 2 | 117 | 104 | |
BC071703.1 | - | 1 - 2 | 123 | 104 | |
DA287424.1 | - | 1 - 2 | 229 | 104 |