Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021927.2 | NP_068746.2 | 11 - 12 | 1538 | 145 |
XM_005248122.3 | XP_005248179.1 | 10 - 11 | 1392 | 145 | |
XM_011513732.2 | XP_011512034.1 | 11 - 12 | 1559 | 145 | |
GenBank mRNA | AK023282.1 | - | 11 - 12 | 1488 | 145 |
AK025248.1 | - | 3 - 4 | 318 | 145 | |
AK225314.1 | - | 11 - 12 | 1493 | 145 | |
BC036768.1 | - | 11 - 12 | 1499 | 145 |