Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021927.2 | NP_068746.2 | 12 - 13 | 1686 | 147 |
XM_005248122.3 | XP_005248179.1 | 11 - 12 | 1540 | 147 | |
XM_011513732.2 | XP_011512034.1 | 12 - 13 | 1707 | 147 | |
GenBank mRNA | AK023282.1 | - | 12 - 13 | 1636 | 147 |
AK025248.1 | - | 4 - 5 | 466 | 147 | |
AK225314.1 | - | 12 - 13 | 1641 | 147 | |
BC036768.1 | - | 12 - 13 | 1647 | 147 |