Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021927.2 | NP_068746.2 | 3 - 4 | 633 | 94 |
XM_005248122.3 | XP_005248179.1 | 2 - 3 | 487 | 94 | |
XM_011513732.2 | XP_011512034.1 | 3 - 4 | 654 | 94 | |
GenBank mRNA | AK023282.1 | - | 3 - 4 | 583 | 94 |
AK225314.1 | - | 3 - 4 | 94 | ||
BC036768.1 | - | 3 - 4 | 594 | 94 |