Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015072.4 | NP_055887.3 | 28 - 29 | 3599 | 76 |
GenBank mRNA | AB023215.1 | - | 26 - 27 | 3159 | 76 |
AL133586.1 | - | 3 - 4 | 480 | 76 | |
AL136808.1 | - | 14 - 15 | 2047 | 76 | |
AY237126.1 | - | 28 - 29 | 3529 | 76 | |
BC136472.1 | - | 28 - 29 | 3489 | 76 | |
BC136473.1 | - | 28 - 29 | 3489 | 76 | |
CR533554.1 | - | 13 - 14 | 1977 | 76 |