Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012093.3 | NP_036225.2 | 12 - 13 | 2372 | 94 |
NM_174858.2 | NP_777283.1 | 12 - 13 | 1761 | 94 | |
XM_005270739.4 | XP_005270796.1 | 12 - 13 | 1608 | 94 | |
XM_006710572.3 | XP_006710635.1 | 12 - 13 | 1539 | 94 | |
XM_017001008.1 | XP_016856497.1 | 12 - 13 | 1556 | 94 | |
XM_017001009.1 | XP_016856498.1 | 12 - 13 | 2264 | 94 | |
XM_017001010.1 | XP_016856499.1 | 12 - 13 | 2591 | 94 | |
XM_017001011.1 | XP_016856500.1 | 12 - 13 | 1374 | 94 | |
GenBank mRNA | AF062595.1 | - | 4 - 5 | 379 | 94 |
AF445193.1 | - | 12 - 13 | 1761 | 94 | |
AY171600.2 | - | 12 - 13 | 2377 | 94 | |
BC012467.2 | - | 7 - 8 | 724 | 94 | |
BC033896.1 | - | 12 - 13 | 2377 | 94 | |
BC036666.1 | - | 12 - 13 | 1687 | 94 | |
CR541890.1 | - | 3 - 4 | 329 | 94 |