Assay Design



Gene Transcripts

Gene Symbol
MDN1
Entrez Gene ID
23195
Gene Name
midasin AAA ATPase 1
Gene Aliases
Rea1
Chromosome Location
Chr.6: 89642775 - 89828021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014611.2 NP_055426.1 26 - 27 3948 70
XM_011535635.2 XP_011533937.1 26 - 27 3753 70
XM_011535636.2 XP_011533938.1 26 - 27 3918 70
GenBank mRNA AF503925.1 - 26 - 27 3877 70

View Full Product Details