Assay Design



Gene Transcripts

Gene Symbol
MDN1
Entrez Gene ID
23195
Gene Name
midasin AAA ATPase 1
Gene Aliases
Rea1
Chromosome Location
Chr.6: 89642775 - 89828021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014611.2 NP_055426.1 40 - 41 131
XM_005248700.3 XP_005248757.1 1 - 2 89 132
XM_006715405.3 XP_006715468.1 4 - 5 131
XM_011535635.2 XP_011533937.1 40 - 41 131
XM_011535636.2 XP_011533938.1 40 - 41 131
GenBank mRNA AF503925.1 - 40 - 41 131

View Full Product Details