Assay Design



Gene Transcripts

Gene Symbol
MDN1
Entrez Gene ID
23195
Gene Name
midasin AAA ATPase 1
Gene Aliases
Rea1
Chromosome Location
Chr.6: 89642775 - 89828021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014611.2 NP_055426.1 67 - 68 11537 115
XM_005248700.3 XP_005248757.1 28 - 29 5469 115
XM_006715405.3 XP_006715468.1 31 - 32 7015 115
XM_011535635.2 XP_011533937.1 67 - 68 11342 115
XM_011535636.2 XP_011533938.1 67 - 68 11507 115
GenBank mRNA AB002299.2 - 21 - 22 4193 115
AF503925.1 - 67 - 68 11466 115
AK125181.1 - 5 - 6 1337 115
CR627399.1 - 13 - 14 2992 115

View Full Product Details