Assay Design



Gene Transcripts

Gene Symbol
IGSF9B
Entrez Gene ID
22997
Gene Name
immunoglobulin superfamily member 9B
Gene Aliases
-
Chromosome Location
Chr.11: 133908564 - 133956960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277285.1 NP_001264214.1 12 - 13 1635 70
XM_011542692.2 XP_011540994.1 13 - 14 2310 70
GenBank mRNA AK097578.1 - 9 - 10 1365 70

View Full Product Details