Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007187.3 | NP_009118.1 | 9 - 10 | 1031 | 119 |
XM_005266245.2 | XP_005266302.1 | 9 - 10 | 1048 | 119 | |
XM_017020380.1 | XP_016875869.1 | 8 - 9 | 961 | 119 | |
GenBank mRNA | AF071185.1 | - | 9 - 10 | 1067 | 119 |
AK000979.1 | - | 9 - 10 | 1084 | 119 | |
AK001557.1 | - | 9 - 10 | 1066 | 119 | |
AK297536.1 | - | 8 - 9 | 969 | 119 | |
AK314525.1 | - | 9 - 10 | 1068 | 119 | |
BC104879.1 | - | 9 - 10 | 975 | 119 | |
BC108310.1 | - | 9 - 10 | 1032 | 119 |