Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136051.2 | NP_001129523.1 | 14 - 15 | 1961 | 82 |
NM_153377.4 | NP_700356.2 | 14 - 15 | 2380 | 82 | |
XM_011537893.2 | XP_011536195.1 | 14 - 15 | 2052 | 82 | |
GenBank mRNA | AK074921.1 | - | 5 - 6 | 880 | 82 |
AK292857.1 | - | 14 - 15 | 2361 | 82 | |
AL833710.1 | - | 3 - 4 | 717 | 82 | |
AY358288.1 | - | 14 - 15 | 2323 | 82 | |
AY358295.1 | - | 14 - 15 | 1961 | 82 | |
AY505340.1 | - | 14 - 15 | 2131 | 82 | |
BC126169.1 | - | 14 - 15 | 2276 | 82 | |
BC126171.1 | - | 14 - 15 | 2276 | 82 | |
BC143960.1 | - | 15 - 16 | 2386 | 82 |