Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153377.4 | NP_700356.2 | 1 - 2 | 537 | 116 |
XM_017018790.1 | XP_016874279.1 | 1 - 2 | 539 | 116 | |
GenBank mRNA | AK292857.1 | - | 1 - 2 | 518 | 116 |
AY358288.1 | - | 1 - 2 | 480 | 116 | |
AY505340.1 | - | 1 - 2 | 288 | 116 | |
BC126169.1 | - | 1 - 2 | 433 | 116 | |
BC126171.1 | - | 1 - 2 | 433 | 116 | |
BC143960.1 | - | 1 - 2 | 433 | 116 |