Assay Design



Gene Transcripts

Gene Symbol
FCRL1
Entrez Gene ID
115350
Gene Name
Fc receptor like 1
Gene Aliases
CD307a, FCRH1, IFGP1, IRTA5
Chromosome Location
Chr.1: 157792249 - 157820150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052938.4 NP_443170.1 10 - 11 1330 78
XM_005244867.2 XP_005244924.1 9 - 10 1184 78
XM_005244869.3 XP_005244926.1 8 - 9 1026 78
XM_011509127.2 XP_011507429.1 9 - 10 1293 78
XM_011509133.2 XP_011507435.1 10 - 11 1155 78
XM_011509134.2 XP_011507436.1 9 - 10 1130 78
XM_011509135.2 XP_011507437.1 9 - 10 1047 78
XM_011509137.2 XP_011507439.1 7 - 8 675 78
XM_017000227.1 XP_016855716.1 9 - 10 3297 78
GenBank mRNA AF459634.1 - 10 - 11 1283 78
AK315748.1 - 10 - 11 1258 78
AY043464.1 - 10 - 11 1285 78
BC021274.2 - 10 - 11 998 78
BC033690.1 - 10 - 11 1281 78

View Full Product Details