Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159397.1 | NP_001152869.1 | 2 - 3 | 167 | 78 |
NM_001159398.1 | NP_001152870.1 | 2 - 3 | 167 | 78 | |
NM_052938.4 | NP_443170.1 | 2 - 3 | 167 | 78 | |
XM_005244866.2 | XP_005244923.1 | 2 - 3 | 138 | 78 | |
XM_005244867.2 | XP_005244924.1 | 2 - 3 | 138 | 78 | |
XM_011509125.1 | XP_011507427.1 | 2 - 3 | 138 | 78 | |
XM_011509126.1 | XP_011507428.1 | 2 - 3 | 138 | 78 | |
XM_011509128.1 | XP_011507430.1 | 2 - 3 | 138 | 78 | |
XM_011509129.1 | XP_011507431.1 | 2 - 3 | 138 | 78 | |
XM_011509130.1 | XP_011507432.1 | 2 - 3 | 138 | 78 | |
XM_011509131.1 | XP_011507433.1 | 2 - 3 | 138 | 78 | |
XM_011509132.2 | XP_011507434.1 | 2 - 3 | 138 | 78 | |
XM_011509136.1 | XP_011507438.1 | 2 - 3 | 138 | 78 | |
GenBank mRNA | AF329488.1 | - | 2 - 3 | 77 | 78 |
AF459634.1 | - | 2 - 3 | 120 | 78 | |
AK096690.1 | - | 2 - 3 | 106 | 78 | |
AK315748.1 | - | 2 - 3 | 95 | 78 | |
AY043464.1 | - | 2 - 3 | 122 | 78 | |
BC021274.2 | - | 2 - 3 | 90 | 78 | |
BC033690.1 | - | 2 - 3 | 118 | 78 |