Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006770.3 | NP_006761.1 | 1 - 2 | 230 | 66 |
XM_011512082.1 | XP_011510384.1 | 1 - 2 | 233 | 66 | |
XM_017005171.1 | XP_016860660.1 | 1 - 2 | 233 | 66 | |
GenBank mRNA | AF035819.1 | - | 1 - 2 | 98 | 66 |
AK297030.1 | - | 1 - 2 | 77 | 66 | |
AK313139.1 | - | 1 - 2 | 179 | 66 | |
AX429246.1 | - | 1 - 2 | 98 | 66 | |
BC016004.1 | - | 1 - 2 | 234 | 66 |