Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032381.1 | NP_001027553.1 | 5 - 6 | 702 | 112 |
NM_001032382.1 | NP_001027554.1 | 5 - 6 | 770 | 112 | |
NM_001032383.1 | NP_001027555.1 | 5 - 6 | 781 | 112 | |
NM_001032384.1 | NP_001027556.1 | 5 - 6 | 690 | 112 | |
NM_001167990.1 | NP_001161462.1 | 5 - 6 | 660 | 112 | |
NM_001167992.1 | NP_001161464.1 | 4 - 5 | 299 | 112 | |
NM_005710.2 | NP_005701.1 | 4 - 5 | 835 | 112 | |
XM_011543884.2 | XP_011542186.1 | 5 - 6 | 840 | 112 | |
GenBank mRNA | AB016533.1 | - | 5 - 6 | 751 | 112 |
AB041833.1 | - | 4 - 5 | 595 | 112 | |
AJ005893.1 | - | 4 - 5 | 838 | 112 | |
AJ242829.1 | - | 5 - 6 | 641 | 112 | |
AJ973594.1 | - | 5 - 6 | 575 | 112 | |
AJ973601.1 | - | 5 - 6 | 683 | 112 | |
AJ973602.1 | - | 4 - 5 | 900 | 112 | |
AJ973605.1 | - | 4 - 5 | 296 | 112 | |
BC012358.1 | - | 5 - 6 | 662 | 112 | |
BX362311.2 | - | 5 - 6 | 739 | 112 | |
CD051018.1 | - | 5 - 6 | 681 | 112 | |
CD365745.1 | - | 2 - 3 | 112 | ||
KU178500.1 | - | 4 - 5 | 581 | 112 | |
KU178501.1 | - | 5 - 6 | 629 | 112 |