Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207354.2 | NP_997237.2 | 8 - 9 | 1051 | 73 |
NR_030767.1 | 9 - 10 | 1134 | 73 | ||
XM_011544985.1 | XP_011543287.1 | 8 - 9 | 994 | 73 | |
XM_011544986.1 | XP_011543288.1 | 9 - 10 | 1134 | 73 | |
XM_011544987.2 | XP_011543289.1 | 8 - 9 | 968 | 73 | |
GenBank mRNA | AK057681.1 | - | 1 - 2 | 1665 | 73 |
AK096277.1 | - | 8 - 9 | 964 | 73 | |
AK124721.1 | - | 9 - 10 | 1131 | 73 | |
AK126438.1 | - | 7 - 8 | 1048 | 73 | |
BC110419.1 | - | 8 - 9 | 1024 | 73 | |
BC121024.1 | - | 8 - 9 | 925 | 73 | |
BC121025.2 | - | 8 - 9 | 925 | 73 |