Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017593.3 | NP_060063.2 | 4 - 5 | 712 | 102 |
NM_198892.1 | NP_942595.1 | 4 - 5 | 712 | 102 | |
XM_005263117.1 | XP_005263174.1 | 4 - 5 | 687 | 102 | |
XM_011532102.1 | XP_011530404.1 | 4 - 5 | 687 | 102 | |
XM_017008381.1 | XP_016863870.1 | 4 - 5 | 464 | 102 | |
XM_017008382.1 | XP_016863871.1 | 2 - 3 | 249 | 102 | |
GenBank mRNA | AK292147.1 | - | 4 - 5 | 688 | 102 |
AL137661.1 | - | 4 - 5 | 748 | 102 | |
BC036021.1 | - | 4 - 5 | 673 | 102 |