Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014976.1 | NP_055791.1 | 32 - 33 | 5047 | 61 |
XM_005269647.3 | XP_005269704.1 | 32 - 33 | 5090 | 61 | |
XM_011539538.2 | XP_011537840.1 | 32 - 33 | 5038 | 61 | |
XM_011539539.2 | XP_011537841.1 | 32 - 33 | 5035 | 61 | |
XM_011539540.1 | XP_011537842.1 | 31 - 32 | 4904 | 61 | |
GenBank mRNA | D80007.1 | - | 32 - 33 | 5000 | 61 |