Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020232.4 | NP_064617.2 | 2 - 3 | 342 | 73 |
NM_147163.1 | NP_671692.1 | 2 - 3 | 200 | 73 | |
GenBank mRNA | AF168712.1 | - | 2 - 3 | 274 | 73 |
AF276707.1 | - | 2 - 3 | 902 | 73 | |
AK057005.1 | - | 2 - 3 | 274 | 73 | |
BC013356.2 | - | 2 - 3 | 265 | 73 | |
BF668035.1 | - | 2 - 3 | 343 | 73 | |
CR457181.1 | - | 1 - 2 | 102 | 73 |