Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033274.4 | NP_150377.1 | 14 - 15 | 1725 | 107 |
XM_005266003.2 | XP_005266060.1 | 14 - 15 | 1677 | 107 | |
XM_011534682.2 | XP_011532984.1 | 7 - 8 | 2481 | 107 | |
XM_017010009.1 | XP_016865498.1 | 7 - 8 | 942 | 107 | |
GenBank mRNA | AF134707.1 | - | 11 - 12 | 1306 | 107 |
AF311317.1 | - | 14 - 15 | 1634 | 107 | |
AF326918.1 | - | 14 - 15 | 1646 | 107 | |
AK091464.1 | - | 14 - 15 | 1687 | 107 | |
AK308182.1 | - | 14 - 15 | 1677 | 107 | |
BC024214.2 | - | 3 - 4 | 310 | 107 | |
BC033132.1 | - | 3 - 4 | 310 | 107 | |
Y13786.2 | - | 14 - 15 | 1675 | 107 |