Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136025.4 | NP_001129497.1 | 11 - 12 | 1399 | 119 |
NM_001172335.2 | NP_001165806.1 | 11 - 12 | 1318 | 119 | |
NM_001282337.1 | NP_001269266.1 | 13 - 14 | 1830 | 119 | |
NM_001282338.1 | NP_001269267.1 | 12 - 13 | 1781 | 119 | |
NM_005032.6 | NP_005023.2 | 11 - 12 | 1684 | 119 | |
XM_011537534.1 | XP_011535836.1 | 11 - 12 | 1375 | 119 | |
GenBank mRNA | AK222801.1 | - | 11 - 12 | 1340 | 119 |
AK294385.1 | - | 3 - 4 | 382 | 119 | |
AK294509.1 | - | 13 - 14 | 1452 | 119 | |
AK295429.1 | - | 2 - 3 | 148 | 119 | |
AK298501.1 | - | 7 - 8 | 586 | 119 | |
AK298530.1 | - | 11 - 12 | 1306 | 119 | |
AK300575.1 | - | 12 - 13 | 1735 | 119 | |
AK311211.1 | - | 6 - 7 | 686 | 119 | |
AK312391.1 | - | 11 - 12 | 1358 | 119 | |
BC008588.1 | - | 5 - 6 | 604 | 119 | |
BC039049.1 | - | 11 - 12 | 1298 | 119 | |
BC056898.1 | - | 11 - 12 | 1402 | 119 | |
M22299.1 | - | 9 - 10 | 1085 | 119 |