Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136025.4 | NP_001129497.1 | 13 - 14 | 1642 | 106 |
NM_001172335.2 | NP_001165806.1 | 13 - 14 | 1561 | 106 | |
NM_001282337.1 | NP_001269266.1 | 15 - 16 | 2073 | 106 | |
NM_001282338.1 | NP_001269267.1 | 14 - 15 | 2024 | 106 | |
NM_005032.6 | NP_005023.2 | 13 - 14 | 1927 | 106 | |
XM_011537534.1 | XP_011535836.1 | 13 - 14 | 1618 | 106 | |
GenBank mRNA | AK222801.1 | - | 13 - 14 | 1583 | 106 |
AK294385.1 | - | 5 - 6 | 625 | 106 | |
AK294509.1 | - | 15 - 16 | 1695 | 106 | |
AK295429.1 | - | 4 - 5 | 391 | 106 | |
AK298530.1 | - | 13 - 14 | 1549 | 106 | |
AK300575.1 | - | 14 - 15 | 1978 | 106 | |
AK312391.1 | - | 13 - 14 | 1601 | 106 | |
BC008588.1 | - | 7 - 8 | 847 | 106 | |
BC039049.1 | - | 13 - 14 | 1541 | 106 | |
BC056898.1 | - | 13 - 14 | 1645 | 106 | |
M22299.1 | - | 11 - 12 | 1328 | 106 |