Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136025.4 | NP_001129497.1 | 3 - 4 | 359 | 98 |
NM_001282337.1 | NP_001269266.1 | 4 - 5 | 763 | 98 | |
NM_001282338.1 | NP_001269267.1 | 4 - 5 | 741 | 98 | |
NM_005032.6 | NP_005023.2 | 3 - 4 | 644 | 98 | |
XM_011537534.1 | XP_011535836.1 | 3 - 4 | 335 | 98 | |
GenBank mRNA | AK294509.1 | - | 4 - 5 | 385 | 98 |
AK298501.1 | - | 3 - 4 | 320 | 98 | |
AK300575.1 | - | 4 - 5 | 695 | 98 | |
AK311397.1 | - | 4 - 5 | 704 | 98 | |
AK312391.1 | - | 3 - 4 | 318 | 98 | |
BC039049.1 | - | 3 - 4 | 258 | 98 | |
BC056898.1 | - | 3 - 4 | 362 | 98 | |
BP214207.1 | - | 3 - 4 | 358 | 98 | |
DC357886.1 | - | 3 - 4 | 359 | 98 | |
M22299.1 | - | 1 - 2 | 45 | 98 | |
M34427.1 | - | 3 - 4 | 312 | 98 |