Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145114.1 | NP_001138586.1 | 1 - 2 | 279 | 83 |
NM_001284337.1 | NP_001271266.1 | 1 - 2 | 279 | 83 | |
NM_015179.3 | NP_055994.2 | 1 - 2 | 279 | 83 | |
XM_011539555.1 | XP_011537857.1 | 1 - 2 | 232 | 83 | |
XM_011539556.2 | XP_011537858.1 | 1 - 2 | 249 | 83 | |
GenBank mRNA | AB014590.1 | - | 1 - 2 | 192 | 83 |
AK022496.1 | - | 1 - 2 | 250 | 83 | |
AK296315.1 | - | 1 - 2 | 143 | 83 | |
BC012745.2 | - | 1 - 2 | 217 | 83 | |
DA227012.1 | - | 1 - 2 | 249 | 83 | |
DC417040.1 | - | 1 - 2 | 279 | 83 |