Assay Design



Gene Transcripts

Gene Symbol
CHD4
Entrez Gene ID
1108
Gene Name
chromodomain helicase DNA binding protein 4
Gene Aliases
CHD-4, Mi-2b, Mi2-BETA
Chromosome Location
Chr.12: 6570082 - 6607439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273.3 NP_001264.2 17 - 18 2868 65
NM_001297553.1 NP_001284482.1 16 - 17 2713 65
XM_005253668.3 XP_005253725.1 17 - 18 2799 65
XM_006718958.1 XP_006719021.1 17 - 18 2874 65
XM_006718959.1 XP_006719022.1 17 - 18 2874 65
XM_006718960.1 XP_006719023.1 17 - 18 2874 65
XM_006718961.2 XP_006719024.1 17 - 18 2799 65
XM_006718962.1 XP_006719025.1 17 - 18 2835 65
XM_017018725.1 XP_016874214.1 17 - 18 2874 65
XM_017018726.1 XP_016874215.1 17 - 18 2874 65
XM_017018727.1 XP_016874216.1 17 - 18 2874 65
XM_017018728.1 XP_016874217.1 17 - 18 2874 65
XM_017018729.1 XP_016874218.1 17 - 18 2799 65
XM_017018730.1 XP_016874219.1 17 - 18 2835 65
XM_017018731.1 XP_016874220.1 17 - 18 2835 65
XM_017018732.1 XP_016874221.1 17 - 18 2760 65
XM_017018733.1 XP_016874222.1 17 - 18 2760 65
XM_017018734.1 XP_016874223.1 17 - 18 2760 65
GenBank mRNA AK128741.1 - 16 - 17 2758 65
BC038596.1 - 17 - 18 2809 65
CR936680.1 - 16 - 17 2621 65
X86691.1 - 16 - 17 2745 65

View Full Product Details