Assay Design



Gene Transcripts

Gene Symbol
CHD4
Entrez Gene ID
1108
Gene Name
chromodomain helicase DNA binding protein 4
Gene Aliases
CHD-4, Mi-2b, Mi2-BETA
Chromosome Location
Chr.12: 6570082 - 6607439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273.3 NP_001264.2 29 - 30 4581 65
NM_001297553.1 NP_001284482.1 28 - 29 4426 65
XM_005253668.3 XP_005253725.1 29 - 30 4512 65
XM_006718958.1 XP_006719021.1 28 - 29 4671 65
XM_006718959.1 XP_006719022.1 29 - 30 4587 65
XM_006718960.1 XP_006719023.1 29 - 30 4584 65
XM_006718961.2 XP_006719024.1 29 - 30 4512 65
XM_006718962.1 XP_006719025.1 29 - 30 4548 65
XM_017018725.1 XP_016874214.1 29 - 30 4587 65
XM_017018726.1 XP_016874215.1 29 - 30 4587 65
XM_017018727.1 XP_016874216.1 29 - 30 4584 65
XM_017018728.1 XP_016874217.1 29 - 30 4584 65
XM_017018729.1 XP_016874218.1 29 - 30 4512 65
XM_017018730.1 XP_016874219.1 29 - 30 4548 65
XM_017018731.1 XP_016874220.1 29 - 30 4548 65
XM_017018732.1 XP_016874221.1 29 - 30 4473 65
XM_017018733.1 XP_016874222.1 29 - 30 4473 65
XM_017018734.1 XP_016874223.1 29 - 30 4473 65
GenBank mRNA AK128741.1 - 28 - 29 4471 65
BC038596.1 - 28 - 29 4606 65
CR936680.1 - 28 - 29 4334 65
X86691.1 - 28 - 29 4458 65

View Full Product Details