Assay Design



Gene Transcripts

Gene Symbol
CHD4
Entrez Gene ID
1108
Gene Name
chromodomain helicase DNA binding protein 4
Gene Aliases
CHD-4, Mi-2b, Mi2-BETA
Chromosome Location
Chr.12: 6570082 - 6607439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273.3 NP_001264.2 37 - 38 5575 91
NM_001297553.1 NP_001284482.1 36 - 37 5420 91
XM_005253668.3 XP_005253725.1 37 - 38 5506 91
XM_006718958.1 XP_006719021.1 36 - 37 5665 91
XM_006718959.1 XP_006719022.1 37 - 38 5587 91
XM_006718960.1 XP_006719023.1 37 - 38 5584 91
XM_006718961.2 XP_006719024.1 37 - 38 5512 91
XM_006718962.1 XP_006719025.1 37 - 38 5548 91
XM_017018725.1 XP_016874214.1 37 - 38 5587 91
XM_017018726.1 XP_016874215.1 37 - 38 5581 91
XM_017018727.1 XP_016874216.1 37 - 38 5578 91
XM_017018728.1 XP_016874217.1 37 - 38 5578 91
XM_017018729.1 XP_016874218.1 37 - 38 5506 91
XM_017018730.1 XP_016874219.1 37 - 38 5542 91
XM_017018731.1 XP_016874220.1 37 - 38 5542 91
XM_017018732.1 XP_016874221.1 37 - 38 5473 91
XM_017018733.1 XP_016874222.1 37 - 38 5467 91
XM_017018734.1 XP_016874223.1 37 - 38 5467 91
GenBank mRNA AK128741.1 - 36 - 37 5465 91
BC038596.1 - 36 - 37 5600 91
CR936680.1 - 36 - 37 5328 91
X86691.1 - 36 - 37 5452 91

View Full Product Details