Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029998.5 | NP_001025169.1 | 1 - 2 | 353 | 138 |
NM_001300842.2 | NP_001287771.1 | 1 - 2 | 353 | 138 | |
NM_001317816.1 | NP_001304745.1 | 1 - 2 | 353 | 138 | |
NM_032128.4 | NP_115504.1 | 1 - 2 | 353 | 138 | |
NR_133924.1 | 1 - 2 | 353 | 138 | ||
XM_011532311.2 | XP_011530613.1 | 1 - 2 | 353 | 138 | |
XM_017008689.1 | XP_016864178.1 | 1 - 2 | 353 | 138 | |
XM_017008690.1 | XP_016864179.1 | 1 - 2 | 353 | 138 | |
XM_017008691.1 | XP_016864180.1 | 1 - 2 | 353 | 138 | |
GenBank mRNA | AK291046.1 | - | 1 - 2 | 221 | 138 |
AL136728.1 | - | 1 - 2 | 281 | 138 | |
AY346324.1 | - | 1 - 2 | 316 | 138 | |
BC063471.1 | - | 1 - 2 | 266 | 138 | |
BC120970.1 | - | 1 - 2 | 212 | 138 | |
BC127626.1 | - | 1 - 2 | 212 | 138 | |
BC127627.1 | - | 1 - 2 | 212 | 138 | |
BC148252.1 | - | 1 - 2 | 212 | 138 | |
BC150308.1 | - | 1 - 2 | 322 | 138 | |
CR933647.1 | - | 1 - 2 | 347 | 138 | |
CR936751.1 | - | 1 - 2 | 210 | 138 | |
DA590625.1 | - | 1 - 2 | 354 | 138 | |
DB205503.1 | - | 1 - 2 | 353 | 138 | |
DQ122860.1 | - | 1 - 2 | 99 | 138 | |
DQ122861.1 | - | 1 - 2 | 99 | 138 | |
DQ871036.1 | - | 1 - 2 | 99 | 138 |